accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOO

CAS RN 1501-05-9

IUPAC Name: 5-oxo-5-phenylpentanoic acid
Synonyms: butanoic acid, 4-benzoyl-
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: SHKWSBAVRQZYLE-UHFFFAOYSA-N
SMILES: OC(=O)CCCC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)