accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOO

CAS RN 15014-25-2

IUPAC Name: 1,3-dibenzyl propanedioate
Synonyms: 1,3-dibenzyl propanedioate
Molecular Weight (g/mol): 284.31
Molecular Formula: C17H16O4
InChi Key: RYFCSKVXWRJEOB-UHFFFAOYSA-N
SMILES: O=C(CC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)