accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 151169-68-5

BOOHHOOH(E/Z)

CAS RN 151169-68-5

IUPAC Name: 3-[4-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Synonyms: 3-[4-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 191.98
Molecular Formula: C9H9BO4
InChi Key: IEMLKNHGGSYOMP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=CC(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results