missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Carboxyvinyl)benzeneboronic acid, 95%, Thermo Scientific™

Product Code. 11368336
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11368336

Brand: Thermo Scientific Alfa Aesar L15586.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151169-68-5
Molecular Formula C9H9BO4
Molecular Weight (g/mol) 191.977
MDL Number MFCD01075704
InChI Key IEMLKNHGGSYOMP-ZZXKWVIFSA-N
Synonym e-3-4-boronophenyl acrylic acid, 4-2-carboxyvinyl phenylboronic acid, 4-boronocinnamic acid, 3-4-boronophenyl acrylic acid, 4-2-carboxyvinyl benzeneboronic acid, 4-carboxyvin-2-yl phenylboronic acid, 4-e-2-carboxyvinyl phenylboronic acid, 4-phenylpropylester boronic acid, 4-e-2-carboxyvinyl benzeneboronic acid, 2e-3-4-dihydroxyboranyl phenyl prop-2-enoic acid
PubChem CID 5288001
IUPAC Name (E)-3-(4-boronophenyl)prop-2-enoic acid
SMILES B(C1=CC=C(C=C1)C=CC(=O)O)(O)O

Specifications

Melting Point ∼190°C to 196°C
Quantity 250 mg
Beilstein 9196727
Formula Weight 191.98
Percent Purity 95%
Chemical Name or Material 4-(2-Carboxyvinyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.