accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

2H2H2H2H2H2H2H2H2H2H

CAS RN 1517-22-2

IUPAC Name: (²H₁₀)phenanthrene
Synonyms: (²H₁₀)phenanthrene
Molecular Weight (g/mol): 188.3
Molecular Formula: C14H10
InChi Key: YNPNZTXNASCQKK-LHNTUAQVSA-N
SMILES: [2H]C1=C([2H])C([2H])=C2C(C([2H])=C([2H])C3=C([2H])C([2H])=C([2H])C([2H])=C23)=C1[2H]
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)