accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COH

CAS RN 1517-69-7

IUPAC Name: 1-phenylethan-1-ol
Synonyms: methylphenyl carbinol fema number 2685 styralyl alcohol 1-phenethyl alcohol 1-fenylethanol fenyl-methylkarbinol α-methylbenzenemethanol α-phenethyl alcohol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: WAPNOHKVXSQRPX-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results