accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15170-57-7

IUPAC Name: platinum(2+) bis(2,4-dioxopentan-3-ide)
Synonyms: platinum(2+) diacetylacetonate
Molecular Weight (g/mol): 393.3
Molecular Formula: C10H14O4Pt
InChi Key: TUOYDQBYTHSWNN-UHFFFAOYSA-N
SMILES: [Pt++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (2)
  • (1)
  • (2)