accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)OClClONHNH2

CAS RN 15253-89-1

IUPAC Name: (2S)-2-(2,4-dichlorophenoxy)propanehydrazide
Synonyms: (2S)-2-(2,4-dichlorophenoxy)propanehydrazide
Molecular Weight (g/mol): 249.09
Molecular Formula: C9H10Cl2N2O2
InChi Key: ZASHHUFHMQDNNE-YFKPBYRVSA-N
SMILES: C[C@H](OC1=CC=C(Cl)C=C1Cl)C(=O)NN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results