accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15282-88-9

PbCH3OCHCH3OCH3OCHCH3O

CAS RN 15282-88-9

IUPAC Name: λ²-lead(2+) bis(2,4-dioxopentan-3-ide)
Synonyms: λ²-lead(2+) diacetylacetonate
Molecular Weight (g/mol): 405.4
Molecular Formula: C10H14O4Pb
InChi Key: VJRBSSRKZYRKTG-UHFFFAOYSA-N
SMILES: [Pb++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results