accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONON(E)

CAS RN 1533-45-5

IUPAC Name: 2-{4-[(1E)-2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl}-1,3-benzoxazole
Synonyms: fluorescent brightener 393
Molecular Weight (g/mol): 414.46
Molecular Formula: C28H18N2O2
InChi Key: ORACIQIJMCYPHQ-MDZDMXLPSA-N
SMILES: O1C2=CC=CC=C2N=C1C1=CC=C(\C=C\C2=CC=C(C=C2)C2=NC3=CC=CC=C3O2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)