Learn More
1,2-Bis(4-(benzo[d]oxazol-2-yl)phenyl)ethene, TRC
Specifications
Specifications
| Chemical Name or Material | 4,4'-Bis(2-benzoxazolyl)stilbene |
| CAS | 1533-45-5 |
| Molecular Formula | C28 H18 N2 O2 |
| Quantity | 2.5 g |
| Synonym | Benzoxazole, 2,2'-(vinylenedi-p-phenylene)bis- (7CI,8CI), 2,2'-(1,2-Ethenediyldi-4,1-phenylene)bis[benzoxazole], 2,2'-(Vinylenedi-p-phenylene)bisbenzoxazole, 4,4'-Bis(2-benzoxazolyl)stilbene, 4,4'-Di(benzoxazol-2-yl)stilbene, Benetex OB 1, C.I. 40674, C.I. Fluorescent Brightener 393, Eastobrite OB 1, Eastobrite OB 3, Fluorescent Brightener 393, OB 1, Optical Brightener 1, TM, TM (UV stabilizer), Uvitex OB 1, Uvitex OB-ONE, p,p'-Bis(2-benzoxazolyl)stilbene |
| InChI Formula | InChI=1S/C28H18N2O2/c1-3-7-25-23(5-1)29-27(31-25)21-15-11-19(12-16-21)9-10-20-13-17-22(18-14-20)28-30-24-6-2-4-8-26(24)32-28/h1-18H |
| SMILES | o1c(nc2ccccc12)c3ccc(C=Cc4ccc(cc4)c5oc6ccccc6n5)cc3 |
| IUPAC Name | 2-[4-[2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-benzoxazole |
| Molecular Weight (g/mol) | 414.45 |
| Formula Weight | 414.1368 |