CAS RN 153559-46-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]benzoic Acid, TRC
CAS: 153559-46-7 Molecular Formula: C23H26O3 Molecular Weight (g/mol): 350.45 Synonym: 4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthyl)carbonyl]benzoic acid, LG 100064, Desmethylene Oxo-bexarotene IUPAC Name: 4-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalene-2-carbonyl)benzoic acid SMILES: Cc1cc2c(cc1C(=O)c3ccc(cc3)C(=O)O)C(C)(C)CCC2(C)C