missing translation for 'onlineSavingsMsg'
Learn More

4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]benzoic Acid, TRC

Product Code. 30420108
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420108 10 mg
30409615 100 mg
30412880 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420108 Supplier Toronto Research Chemicals Supplier No. TRCT29494510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153559-46-7
Molecular Formula C23H26O3
Molecular Weight (g/mol) 350.45
Synonym 4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthyl)carbonyl]benzoic acid, LG 100064, Desmethylene Oxo-bexarotene
IUPAC Name 4-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalene-2-carbonyl)benzoic acid
SMILES Cc1cc2c(cc1C(=O)c3ccc(cc3)C(=O)O)C(C)(C)CCC2(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H26O3/c1-14-12-18-19(23(4,5)11-10-22(18,2)3)13-17(14)20(24)15-6-8-16(9-7-15)21(25)26/h6-9,12-13H,10-11H2,1-5H3,(H,25,26)
Formula Weight 350.1882
Chemical Name or Material 4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.