CAS RN 154154-90-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Deethyl Dorzolamide, TRC
CAS: 154154-90-2 Molecular Formula: C8 H12 N2 O4 S3 Molecular Weight (g/mol): 296.3869 Synonym: 4H-Thieno[2,3-b]thiopyran-2-sulfonamide, 4-amino-5,6-dihydro-6-methyl-, 7,7-dioxide, (4S-trans)- (9CI),N-Deethyldorzolamide,Dorzolamide Related Compound D (USP),(4S-trans)-4-Amino-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide,USP Dorzolamide Related Compound D,Dorzolamide EP Impurity D IUPAC Name: (4S,6S)-4-amino-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide SMILES: C[C@H]1C[C@H](N)c2cc(sc2S1(=O)=O)S(=O)(=O)N