missing translation for 'onlineSavingsMsg'
Learn More

N-Deethyl Dorzolamide, TRC

Product Code. 30134425
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134425 100 mg 100mg
30005994 5 mg 5mg
30010575 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134425 Supplier Toronto Research Chemicals Supplier No. TRCD228550100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154154-90-2
Molecular Formula C8 H12 N2 O4 S3
Molecular Weight (g/mol) 296.3869
Synonym 4H-Thieno[2,3-b]thiopyran-2-sulfonamide, 4-amino-5,6-dihydro-6-methyl-, 7,7-dioxide, (4S-trans)- (9CI), N-Deethyldorzolamide, Dorzolamide Related Compound D (USP), (4S-trans)-4-Amino-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide, USP Dorzolamide Related Compound D, Dorzolamide EP Impurity D
IUPAC Name (4S,6S)-4-amino-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide
SMILES C[C@H]1C[C@H](N)c2cc(sc2S1(=O)=O)S(=O)(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H12N2O4S3/c1-4-2-6(9)5-3-7(17(10,13)14)15-8(5)16(4,11)12/h3-4,6H,2,9H2,1H3,(H2,10,13,14)/t4-,6-/m0/s1
Formula Weight 295.9959
Chemical Name or Material N-Deethyl Dorzolamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.