CAS RN 154447-36-6
IUPAC Name:
2-(morpholin-4-yl)-8-phenyl-4H-chromen-4-one
Synonyms:
2-(morpholin-4-yl)-8-phenylchromen-4-one
Molecular Weight (g/mol):
307.35
Molecular Formula:
C19H17NO3
InChi Key:
CZQHHVNHHHRRDU-UHFFFAOYSA-N
SMILES:
O=C1C=C(OC2=C(C=CC=C12)C1=CC=CC=C1)N1CCOCC1
1
–
3
of
3
results
1
LY 294002, TRC
CAS: 154447-36-6 Molecular Formula: C19H17NO3 Molecular Weight (g/mol): 307.34 Synonym: NSC 697286,SF 1101,8-Phenyl-2-(morpholin-4-yl)-chromen-4-one,2-(4-Morpholinyl)-8-phenyl-4H-1-benzopyran-4-one,2-Morpholino-8-phenyl-4H-chromen-4-one SMILES: COC(=O)C[C@@H](Br)C(=O)OC
3