missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-8-phenyl-4H-chromen-4-one, 98%, BLD Pharm™

Product Code. 30780408
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30780408 100 mg 100mg
30773216 50 mg 50mg
30772809 250 mg 250mg
30774769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30780408 Supplier BLD Pharm Supplier No. BD136896100mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 154447-36-6
Molecular Formula C19H17NO3
MDL Number MFCD00270881
Quantity 100 mg
SMILES O=C1C=C(N2CCOCC2)OC3=C1C=CC=C3C4=CC=CC=C4
Molecular Weight (g/mol) 307.34
Percent Purity 98%
Chemical Name or Material 2-Morpholino-8-phenyl-4H-chromen-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.