accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOBr(E/Z)

CAS RN 154561-38-3

IUPAC Name: 2-oxo-2-phenylethyl 4-bromobut-2-enoate
Synonyms: 2-oxo-2-phenylethyl 4-bromobut-2-enoate
Molecular Weight (g/mol): 283.12
Molecular Formula: C12H11BrO3
InChi Key: JARTYZWEUJDLPS-UHFFFAOYSA-N
SMILES: BrCC=CC(=O)OCC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results