CAS RN 15492-51-0
IUPAC Name:
terbium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Synonyms:
terbium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Molecular Weight (g/mol):
708.74
Molecular Formula:
C33H57O6Tb
InChi Key:
JSAHJYGCYOFNGW-UHFFFAOYSA-K
SMILES:
[Tb+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
1
–
1
of
1
results
1
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)terbium(III), 99%, Thermo Scientific Chemicals
CAS: 15492-51-0 Molecular Formula: C33H57O6Tb Molecular Weight (g/mol): 708.74 MDL Number: MFCD00064752 InChI Key: DIIXZXQKWUIOAN-UHFFFAOYSA-N Synonym: tb tmhd 3,terbium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate PubChem CID: 133109942 SMILES: [Tb+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C