accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15545-48-9

IUPAC Name: 1-(3-chloro-4-methylphenyl)-3,3-dimethylurea
Synonyms: chlortoluron chlorotoluron
Molecular Weight (g/mol): 212.68
Molecular Formula: C10H13ClN2O
InChi Key: JXCGFZXSOMJFOA-UHFFFAOYSA-N
SMILES: CN(C)C(=O)NC1=CC=C(C)C(Cl)=C1
Concentration 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results