missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Chlortoluron in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12952534 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12952534 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12952534 Supplier Fisher Chemical Supplier No. P812950

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 15545-48-9
Molecular Formula C10H13ClN2O
Concentration 100 μg/mL
MDL Number MFCD00018275
InChI Key JXCGFZXSOMJFOA-UHFFFAOYSA-N
SMILES CN(C)C(=O)NC1=CC=C(C)C(Cl)=C1
IUPAC Name 1-(3-chloro-4-methylphenyl)-3,3-dimethylurea
Solvent or Matrix Methanol
Molecular Weight (g/mol) 212.68
PubChem CID 27375
Quantity 1 mL
ChEBI CHEBI:81981
Chemical Name or Material Chlortoluron
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.