CAS RN 155603-50-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mono(2-ethylhexyl) Terephthalate, TRC
CAS: 155603-50-2 Molecular Formula: C16 H22 O4 Molecular Weight (g/mol): 278.34 Synonym: 1,4-Benzenedicarboxylic acid, mono(2-ethylhexyl) ester (9CI),2-Ethylhexyl terephthalate,Mono(2-ethylhexyl) terephthalate,Mono(2-Ethylhexyl) Terephthalate,Terephthalic acid, mono-2-ethylhexyl ester IUPAC Name: 4-(2-ethylhexoxycarbonyl)benzoic acid SMILES: CCCCC(CC)COC(=O)c1ccc(cc1)C(=O)O