CAS RN 15573-38-3
IUPAC Name:
tris(trimethylsilyl)phosphane
Synonyms:
tris(trimethylsilyl)phosphane
Molecular Weight (g/mol):
250.54
Molecular Formula:
C9H27PSi3
InChi Key:
OUMZKMRZMVDEOF-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C
1
–
1
of
1
results
1
[Tris(trimethylsilyl)]phosphine, 98%, Thermo Scientific Chemicals
CAS: 15573-38-3 Molecular Formula: C9H27PSi3 MDL Number: MFCD00015487 InChI Key: OUMZKMRZMVDEOF-UHFFFAOYSA-N Synonym: tris trimethylsilyl phosphine,tris trimethylsilyl phosphane,phosphine, tris trimethylsilyl,1,1,1,3,3,3-hexamethyl-2-trimethylsilyl disilaphosphane,tris trimethylsilyl phosphine in hexane,pubchem6456,acmc-1btki,tris-trimethylsilyl phosphine,phosphine,tris trimethylsilyl PubChem CID: 272683 IUPAC Name: tris(trimethylsilyl)phosphane SMILES: C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C