missing translation for 'onlineSavingsMsg'
Learn More

[Tris(trimethylsilyl)]phosphine, 98%, Thermo Scientific Chemicals

Product Code. 10666281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10666281 5 mL 5mL
10547231 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10666281 Supplier Thermo Scientific Acros Supplier No. 291700050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15573-38-3
Molecular Formula C9H27PSi3
MDL Number MFCD00015487
InChI Key OUMZKMRZMVDEOF-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphine, tris trimethylsilyl phosphine in hexane, acmc-1btki, phosphine,tris trimethylsilyl, tris-trimethylsilyl phosphine, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl disilaphosphane, phosphine, tris trimethylsilyl, pubchem6456, tris trimethylsilyl phosphane
PubChem CID 272683
IUPAC Name tris(trimethylsilyl)phosphane
SMILES C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.86 g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [(CH3)3Si]3P
Quantity 5 mL
Molecular Weight (g/mol) 250.54
Formula Weight 250.54
Physical Form Liquid
Chemical Name or Material [Tris(trimethylsilyl)]phosphine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.