CAS RN 155742-64-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methylcarbonyl-2-chloroacetamidrazone, TRC
CAS: 155742-64-6 Molecular Formula: C4 H8 Cl N3 O2 Molecular Weight (g/mol): 165.58 IUPAC Name: methyl N-[(2-chloroethanimidoyl)amino]carbamate SMILES: COC(=O)NNC(=N)CCl