missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcarbonyl-2-chloroacetamidrazone, TRC

Product Code. 30004148
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004148 100 mg 100mg
30004145 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004148 Supplier Toronto Research Chemicals Supplier No. TRCM261290100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155742-64-6
Molecular Formula C4 H8 Cl N3 O2
Molecular Weight (g/mol) 165.58
IUPAC Name methyl N-[(2-chloroethanimidoyl)amino]carbamate
SMILES COC(=O)NNC(=N)CCl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H8ClN3O2/c1-10-4(9)8-7-3(6)2-5/h2H2,1H3,(H2,6,7)(H,8,9)
Formula Weight 165.0305
Chemical Name or Material N-Methylcarbonyl-2-chloroacetamidrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.