accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1575-61-7

ClClO

CAS RN 1575-61-7

IUPAC Name: 5-chloropentanoyl chloride
Synonyms: valeryl chloride, 5-chloro-
Molecular Weight (g/mol): 155.02
Molecular Formula: C5H8Cl2O
InChi Key: SVNNWKWHLOJLOK-UHFFFAOYSA-N
SMILES: ClCCCCC(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)