missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleryl Chloride, TRC

Product Code. 30359245
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.50g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359245 500 mg 500mg
30352230 250 mg 250mg
30356362 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359245 Supplier Toronto Research Chemicals Supplier No. TRCC382545500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1575-61-7
Molecular Formula C5H8OCl2
Molecular Weight (g/mol) 155.02
Synonym Apixaban Impurity 82
IUPAC Name 5-chloropentanoyl chloride
SMILES ClCCCCC(=O)Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H8Cl2O/c6-4-2-1-3-5(7)8/h1-4H2
Formula Weight 153.9952
Chemical Name or Material 5-Chlorovaleryl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.