accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1588508-08-0

CH3OFFFOOH(E/Z)

CAS RN 1588508-08-0

IUPAC Name: 3-[2-methyl-4-(trifluoromethoxy)phenyl]prop-2-enoic acid
Synonyms: 3-[2-methyl-4-(trifluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 246.19
Molecular Formula: C11H9F3O3
InChi Key: JYTHJDZEVRTJAK-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(O)=O)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results