accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1593-08-4

IUPAC Name: quinoxaline-2-carbaldehyde
Synonyms: quinoxaline-2-carbaldehyde
Molecular Weight (g/mol): 158.16
Molecular Formula: C9H6N2O
InChi Key: UJEHWLFUEQHEEZ-UHFFFAOYSA-N
SMILES: O=CC1=CN=C2C=CC=CC2=N1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results