accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO(S)O(S)(R)O(R)ONOO

CAS RN 16051-77-7

IUPAC Name: (3R,3aS,6S,6aR)-6-hydroxy-hexahydrofuro[3,2-b]furan-3-yl nitrate
Synonyms: isosorbide mononitrate ismn apogepha ismn heumann monosorbitrate pentacard monis monit elantan long monicor monopront
Molecular Weight (g/mol): 191.14
Molecular Formula: C6H9NO6
InChi Key: YWXYYJSYQOXTPL-SLPGGIOYSA-N
SMILES: O[C@H]1CO[C@@H]2[C@@H](CO[C@H]12)O[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)