missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide mononitrate, 98+%

Product Code. 15415859
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415859 25 g 25g
15405859 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415859 Supplier Thermo Scientific Alfa Aesar Supplier No. J63674.14
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A coronary vasodilator that does not require hepatic biotransformation | CAS: 16051-77-7 | C6H9NO6 | 191.139 g/mol

A coronary vasodilator that does not require hepatic biotransformation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16051-77-7
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.139
MDL Number MFCD00143462
InChI Key YWXYYJSYQOXTPL-SLPGGIOYSA-N
Synonym isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
PubChem CID 27661
ChEBI CHEBI:6062
IUPAC Name [(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
SMILES C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O

Specifications

Melting Point 92°C
Color White
Quantity 25 g
UN Number UN3251
Merck Index 14,5225
Formula Weight 191.14
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Isosorbide mononitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.