accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNSOOOHOH

CAS RN 16052-06-5

IUPAC Name: 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
Synonyms: EPPS hepps (molecule) hepps
Molecular Weight (g/mol): 252.33
Molecular Formula: C9H20N2O4S
InChi Key: OWXMKDGYPWMGEB-UHFFFAOYSA-N
SMILES: OCCN1CCN(CCCS(O)(=O)=O)CC1
Quantity 
  • (1)
  • (5)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
pH 
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (2)
  • (9)
  • (1)
  • (5)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (7)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)