accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1611-56-9

HOBr

CAS RN 1611-56-9

IUPAC Name: 11-bromoundecan-1-ol
Synonyms: 11-bromoundecan-1-ol
Molecular Weight (g/mol): 251.21
Molecular Formula: C11H23BrO
InChi Key: XFGANBYCJWQYBI-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)