CAS RN 16156-94-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Metronidazole EP Impurity F, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
O-Hydroxyethyl Metronidazole, TRC
CAS: 16156-94-8 Molecular Formula: C8 H13 N3 O4 Molecular Weight (g/mol): 215.21 Synonym: 2-[2-(2-methyl-5-nitroimidazol-1-yl)ethoxy]ethanol, Imidazole, 1-((2-hydroxyethoxy)ethyl)-2-methyl-5-nitro IUPAC Name: 2-[2-(2-methyl-5-nitroimidazol-1-yl)ethoxy]ethanol SMILES: Cc1ncc([N+](=O)[O-])n1CCOCCO