missing translation for 'onlineSavingsMsg'
Learn More

O-Hydroxyethyl Metronidazole, TRC

Product Code. 30138491
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138491 50 mg 50mg
30132927 25 mg 25mg
17915045 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138491 Supplier Toronto Research Chemicals Supplier No. TRCH94228050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16156-94-8
Molecular Formula C8 H13 N3 O4
Molecular Weight (g/mol) 215.21
Synonym 2-[2-(2-methyl-5-nitroimidazol-1-yl)ethoxy]ethanol, Imidazole, 1-((2-hydroxyethoxy)ethyl)-2-methyl-5-nitro
IUPAC Name 2-[2-(2-methyl-5-nitroimidazol-1-yl)ethoxy]ethanol
SMILES Cc1ncc([N+](=O)[O-])n1CCOCCO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H13N3O4/c1-7-9-6-8(11(13)14)10(7)2-4-15-5-3-12/h6,12H,2-5H2,1H3
Formula Weight 215.0906
Chemical Name or Material O-Hydroxyethyl Metronidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.