accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONBOHHO

CAS RN 163105-90-6

IUPAC Name: (2-methoxypyridin-3-yl)boronic acid
Synonyms: 2-methoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 152.94
Molecular Formula: C6H8BNO3
InChi Key: NVOLYUXUHWBCRJ-UHFFFAOYSA-N
SMILES: COC1=NC=CC=C1B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results