CAS RN 163515-14-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Dimethenamide-P, TRC
CAS: 163515-14-8 Molecular Formula: C12 H18 Cl N O2 S Molecular Weight (g/mol): 275.79 Synonym: Acetamide, 2-chloro-N-(2,4-dimethyl-3-thienyl)-N-(2-methoxy-1-methylethyl)-, (S)-,2-Chloro-N-(2,4-dimethyl-3-thienyl)-N-[(1S)-2-methoxy-1-methylethyl]acetamide,(S)-Dimethenamid,BAS 656H,DMTA-P,Dimethenamid P,Establish,Frontier X 2,Isard,Outlook,Spectrum,Spectrum (herbicide),Tower (herbicide) IUPAC Name: 2-chloro-N-(2,4-dimethylthiophen-3-yl)-N-[(2S)-1-methoxypropan-2-yl]acetamide SMILES: COC[C@H](C)N(C(=O)CCl)c1c(C)csc1C