accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FBHOOH

CAS RN 163517-61-1

IUPAC Name: (3-fluoro-2-methylphenyl)boronic acid
Synonyms: 3-fluoro-2-methylphenylboronic acid
Molecular Weight (g/mol): 153.95
Molecular Formula: C7H8BFO2
InChi Key: NYBIUWJUWTUGFV-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results