missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15489487
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489487 1 g 1g
15499487 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489487 Supplier Thermo Scientific Alfa Aesar Supplier No. H52752.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163517-61-1
Molecular Formula C7H8BFO2
Molecular Weight (g/mol) 153.947
MDL Number MFCD07368679
InChI Key NYBIUWJUWTUGFV-UHFFFAOYSA-N
Synonym 3-fluoro-2-methylphenyl boronic acid, 2-methyl-3-fluoro-phenylboronic acid, 3-fluoro-2-methylbenzeneboronic acid, 3-fluoro-o-tolylboronic acid, boronic acid, 3-fluoro-2-methylphenyl, boronic acid,b-3-fluoro-2-methylphenyl, 3-fluoro-2-methyl-phenyl boronic acid, 2-borono-6-fluorotoluene, acmc-1c5qk, 2-methyl-3-fluorophenylboronic acid
PubChem CID 2783191
IUPAC Name (3-fluoro-2-methylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)F)C)(O)O

Specifications

Melting Point 142°C to 144°C
Quantity 1 g
Formula Weight 153.95
Percent Purity 98%
Chemical Name or Material 3-Fluoro-2-methylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.