accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NONHNH2N

CAS RN 16352-06-0

IUPAC Name: 4-amino-6-methyl-2,5-dihydro-1,3,5-triazin-2-one
Synonyms: 4-amino-6-methyl-5H-1,3,5-triazin-2-one
Molecular Weight (g/mol): 126.12
Molecular Formula: C4H6N4O
InChi Key: UUTHDVPZNWJUFV-UHFFFAOYSA-N
SMILES: CC1=NC(=O)N=C(N)N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results