missing translation for 'onlineSavingsMsg'
Learn More

Acetoguanide, TRC

Product Code. 30134900
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134900 10 g 10g
17914205 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134900 Supplier Toronto Research Chemicals Supplier No. TRCA16262010G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16352-06-0
Molecular Formula C4 H6 N4 O
Molecular Weight (g/mol) 126.12
Synonym 1,3,5-Triazin-2(1H)-one, 4-amino-6-methyl- (9CI), s-Triazin-2-ol, 4-amino-6-methyl- (6CI,7CI,8CI), 6-Amino-4-methyl-1,3,5-triazin-2(1H)-one, 2-Amino-6-methyl-1H-1,3,5-triazin-4-one, 4-Amino-6-methyl-1,3,5-triazin-2-ol, Acetoguanide, IN-B 5528, 1,3,5-Triazin-2(1H)-one, 6-amino-4-methyl-, 1,3,5-Triazin-2(1H)-one, 4-amino-6-methyl- (9CI), s-Triazin-2-ol, 4-amino-6-methyl- (6CI,7CI,8CI), 6-Amino-4-methyl-1,3,5-triazin-2(1H)-one, 2-Amino-6-methyl-1H-1,3,5-triazin-4-one, 4-Amino-6-methyl-1,3,5-triazin-2-ol, Acetoguanide, IN-B 5528, 1,3,5-Triazin-2(1H)-one, 6-amino-4-methyl-
IUPAC Name 4-amino-6-methyl-1,3,5-triazin-2-ol
SMILES Cc1nc(N)nc(O)n1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H6N4O/c1-2-6-3(5)8-4(9)7-2/h1H3,(H3,5,6,7,8,9)
Formula Weight 126.0542
Chemical Name or Material Acetoguanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.