CAS RN 1637-75-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
m-Hydroxyhippuric Acid, TRC
CAS: 1637-75-8 Molecular Formula: C9H9NO4 Molecular Weight (g/mol): 195.17 Synonym: N-(3-Hydroxybenzoyl)-glycine,3-Hydroxybenzoylglycine,3-Hydroxyhippuric acid,N-m-Hydroxylbenzoylglycine IUPAC Name: 2-[(3-hydroxybenzoyl)amino]acetic acid SMILES: Cl.N#CCCNC(C)Cc1ccccc1