missing translation for 'onlineSavingsMsg'
Learn More

m-Hydroxyhippuric Acid, TRC

Product Code. 30136355
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136355 2.5 g 2.5g
30010247 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136355 Supplier Toronto Research Chemicals Supplier No. TRCH9431252.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-75-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
Synonym N-(3-Hydroxybenzoyl)-glycine, 3-Hydroxybenzoylglycine, 3-Hydroxyhippuric acid, N-m-Hydroxylbenzoylglycine
IUPAC Name 2-[(3-hydroxybenzoyl)amino]acetic acid
SMILES Cl.N#CCCNC(C)Cc1ccccc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H9NO4/c11-7-3-1-2-6(4-7)9(14)10-5-8(12)13/h1-4,11H,5H2,(H,10,14)(H,12,13)
Formula Weight 195.0532
Chemical Name or Material m-Hydroxyhippuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.