CAS RN 163727-74-0
IUPAC Name:
tributyl({4-[2-(N',N''-dicyclohexylcarbamimidamido)-3-(naphthalen-2-yl)propanamido]phenyl}methyl)phosphanium hydrochloride chloride
Synonyms:
tributyl({4-[2-(N',N''-dicyclohexylcarbamimidamido)-3-(naphthalen-2-yl)propanamido]phenyl}methyl)phosphanium hydrochloride chloride
Molecular Weight (g/mol):
783.95
Molecular Formula:
C45H69Cl2N4OP
InChi Key:
YYJGBEZPVOUBMJ-UHFFFAOYNA-N
SMILES:
Cl.[Cl-].CCCC[P+](CCCC)(CCCC)CC1=CC=C(NC(=O)C(CC2=CC=C3C=CC=CC3=C2)NC(NC2CCCCC2)=NC2CCCCC2)C=C1
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ WIN 64338 hydrochloride
Tocris Bioscience™ WIN 64338 hydrochloride
CAS: 163727-74-0 Molecular Formula: C45H69Cl2N4OP