missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WIN 64338 hydrochloride
GREENER_CHOICE

Product Code. 18064162
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064162 10 mg 10mg
18005252 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18064162 Supplier Tocris Bioscience™ Supplier No. 1057/10
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Bradykinin B2 antagonist

The first potent, non-peptide, competitive bradykinin B2 receptor antagonist. In organ bath studies, WIN 64338 inhibits [3H]-bradykinin binding on guinea pig trachea with nanomolar affinity but is not active in the rabbit aorta (the classical bradykinin B1 preparation).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163727-74-0
Molecular Formula C45H69Cl2N4OP

Specifications

Quantity 10 mg
Formula Weight 783.95
Purity >98%
Molecular Weight (g/mol) 783.948
Chemical Name or Material WIN 64338 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.