CAS RN 16474-43-4
CAS RN 16474-43-4
IUPAC Name:
tert-butyl 2,2-dimethylpropanoate
Synonyms:
tert-butyl 2,2-dimethylpropanoate
Molecular Weight (g/mol):
158.24
Molecular Formula:
C9H18O2
InChi Key:
VXHFNALHLRWIIU-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)C(C)(C)C
1
–
2
of
2
results
1
tert-Butyl pivalate, 98%
CAS: 16474-43-4 Molecular Formula: C9H18O2 Molecular Weight (g/mol): 158.24 MDL Number: MFCD01861974 InChI Key: VXHFNALHLRWIIU-UHFFFAOYSA-N PubChem CID: 519272 IUPAC Name: tert-butyl 2,2-dimethylpropanoate SMILES: CC(C)(C)OC(=O)C(C)(C)C