Learn More
tert-Butyl pivalate, 98%
Product Code. 15490158
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490158 5 g 5g
15400168 25 g 25g
15410168 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15490158 Supplier Thermo Scientific Alfa Aesar Supplier No. H53395.06
Request Bulk or Custom Format
CAS: 16474-43-4 | C9H18O2 | 158.24 g/mol
tert-Butyl pivalate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl pivalate is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16474-43-4
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD01861974
InChI Key VXHFNALHLRWIIU-UHFFFAOYSA-N
PubChem CID 519272
IUPAC Name tert-butyl 2,2-dimethylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)C
Specifications
Density 0.813
Boiling Point 134°C to 135°C
Flash Point >62°C (144°F)
Linear Formula (CH3)3CCO2C(CH3)3
Refractive Index 1.392
Quantity 5 g
Synonym tert-butyl pivalate, tert-butyl trimethylacetate, propanoic acid, 2,2-dimethyl, 1,1-dimethylethyl ester, di-tert-butylcarboxylic acid, 2,2-dimethylpropanoic acid tert-butyl ester, 2,2-dimethyl-propionic acid tert-butyl ester, propanoic acid, 2,2-dimethyl-, 1,1-dimethylethyl ester, propanoic acid,2,2-dimethyl-, 1,1-dimethylethyl ester
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material tert-Butyl pivalate