accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16518-62-0

IUPAC Name: 3-bromo-N,N-dimethylaniline
Synonyms: 3-bromo-N,N-dimethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: USEXQPWLCGBYNT-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results