missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-N,N-dimethylaniline, 97%

Product Code. 11374016
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374016 25 g 25g
11364016 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374016 Supplier Thermo Scientific Alfa Aesar Supplier No. L17279.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16518-62-0 | C8H10BrN | 200.08 g/mol

3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2′,4′,6′-tri-i-propyl-1,1′-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.

Solubility
Miscible with chloroform, dichloromethane and methanol.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16518-62-0
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00045020
InChI Key USEXQPWLCGBYNT-UHFFFAOYSA-N
Synonym benzenamine, 3-bromo-n,n-dimethyl, 3-bromophenyl dimethylamine, n,n-dimethyl 3-bromoaniline, 3-bromo-n,n-dimethylbenzenamine, n,n-dimethyl-3-bromoaniline, m-bromodimethyl aniline, 3-bromo-dimethylaniline, m-bromodimethylaniline, pubchem3760, acmc-1c80w
PubChem CID 140102
IUPAC Name 3-bromo-N,N-dimethylaniline
SMILES CN(C)C1=CC=CC(Br)=C1

Specifications

Melting Point 11°C
Density 1.365
Boiling Point 126°C (14 mmHg)
Refractive Index 1.6015
Quantity 25 g
Beilstein 2079908
Solubility Information Miscible with chloroform,dichloromethane and methanol.
Formula Weight 200.08
Percent Purity 97%
Chemical Name or Material 3-Bromo-N,N-dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.